Learn More
Abraxis AbraMag® anti-Human Magnetic Beads, 1 mL Sample Size, 5 mg/mL, Magnetic Beads for Purification
SDP
Catalog No. NC2940199
Encompass
AbraMag® Anti-Human IgG Magnetic Beads are densely coated with Goat anti-Human IgG. They are utilized in the magnetic separation and isolation of human antibodies from serum or human antibody-labeled components.The magnetic beads are incubated with a sample containing antibodies and then held by a magnet next to the tube. After the unbound contaminants are washed from the beads, the bound antibodies are eluted from the beads with an elution buffer (not provided). With the magnet immobilizing the beads, the eluted antibodies are transferred to a new tube.AbraMag® Magnetic Beads are superparamagnetic, non-aggregating iron oxide particles for capturing or purifying targets such as proteins, antigens, antibodies, DNA/RNA, cells, and more. AbraMag® Magnetic Bead’s design enables faster binding kinetics, with high sensitivity & selectivity, in many biomedical and research applications—including automated HTS and manual applications.
Catalog No. NC2940199 Supplier Abraxis Supplier No. 544062
AbraMag® Anti-Human IgG Magnetic Beads are densely coated with Goat anti-Human IgG. They are utilized in the magnetic separation and isolation of human antibodies from serum or human antibody-labeled components.The magnetic beads are incubated with a sample containing antibodies and then held by a magnet next to the tube. After the unbound contaminants are washed from the beads, the bound antibodies are eluted from the beads with an elution buffer (not provided). With the magnet immobilizing the beads, the eluted antibodies are transferred to a new tube.AbraMag® Magnetic Beads are superparamagnetic, non-aggregating iron oxide particles for capturing or purifying targets such as proteins, antigens, antibodies, DNA/RNA, cells, and more. AbraMag® Magnetic Bead’s design enables faster binding kinetics, with high sensitivity & selectivity, in many biomedical and research applications—including automated HTS and manual applications.