Learn More
Electron Microscopy Sciences Flexible Shaft Machine Abrasive Band Fine, 1/2"
SDP
Catalog No. 5024385
Encompass_Preferred
Abrasive bands are glue bonded, silicon carbide for fast cutting. All are 1/2" (12.7 mm) long.
Diameter: 1/2"
Catalog No. 50-243-85 Supplier Electron Microscopy Sciences Supplier No. 6215030
Abrasive bands are glue bonded, silicon carbide for fast cutting. All are 1/2" (12.7 mm) long.
Diameter: 1/2"