Learn More
Electron Microscopy Sciences Flexible Shaft Machine Abrasive Band Fine Grit, 3/8"
SDP
Catalog No. 5024379
Encompass_Preferred
Abrasive bands are glue bonded, silicon carbide for fast cutting. All are 1/2" (12.7 mm) long.
Diameter: 3/8"
Catalog No. 50-243-79 Supplier Electron Microscopy Sciences Supplier No. 6215020
Abrasive bands are glue bonded, silicon carbide for fast cutting. All are 1/2" (12.7 mm) long.
Diameter: 3/8"