Learn More
Electron Microscopy Sciences Flexible Shaft Machine Abrasive Band Medium, 1/2"
SDP
Catalog No. 5024380
Encompass_Preferred
Abrasive bands are glue bonded, silicon carbide for fast cutting. All are 1/2" (12.7 mm) long.
Diameter: 1/2"
Catalog No. 50-243-80 Supplier Electron Microscopy Sciences Supplier No. 6215031
Abrasive bands are glue bonded, silicon carbide for fast cutting. All are 1/2" (12.7 mm) long.
Diameter: 1/2"