Learn More
Electron Microscopy Sciences Flexible Shaft Machine Abrasive Band Medium Grit, 1/4"
SDP
Catalog No. 5024376
Encompass_Preferred
Abrasive bands are glue bonded, silicon carbide for fast cutting. All are 1/2" (12.7 mm) long.
Diameter: 1/4"
Catalog No. 50-243-76 Supplier Electron Microscopy Sciences Supplier No. 6215011
Abrasive bands are glue bonded, silicon carbide for fast cutting. All are 1/2" (12.7 mm) long.
Diameter: 1/4"