Learn More
Grainger 3M Trizact Hookit Foam Abrasive Disc, Silicon Carbide, 150 mm, P3000, 60/Pk.
Catalog No. 502787298
Encompass_Preferred
This structured foam abrasive disc is engineered for professional paint refinishing applications. It features precise microreplicated mineral structures that deliver a consistent, high-quality finish while providing a long-lasting abrasive performance. Designed for wet use, this disc effectively refines 1500 and 2000 grit sand scratches to reduce compounding time.
  • Patterned, precise mineral structures provide a combination of fast cut and consistent finish.
  • Flexible foam backing offers excellent conformability on complex shapes and surfaces.
  • Hookit™ attachment system allows for quick and easy disc changes.
  • Optimized for wet sanding to extend abrasive life and minimize loading.
  • Helps reduce or eliminate full stages in paint finishing processes.
Catalog No. 50-278-7298 Supplier Grainger Supplier No. 191L93
This structured foam abrasive disc is engineered for professional paint refinishing applications. It features precise microreplicated mineral structures that deliver a consistent, high-quality finish while providing a long-lasting abrasive performance. Designed for wet use, this disc effectively refines 1500 and 2000 grit sand scratches to reduce compounding time.
  • Patterned, precise mineral structures provide a combination of fast cut and consistent finish.
  • Flexible foam backing offers excellent conformability on complex shapes and surfaces.
  • Hookit™ attachment system allows for quick and easy disc changes.
  • Optimized for wet sanding to extend abrasive life and minimize loading.
  • Helps reduce or eliminate full stages in paint finishing processes.