Learn More
Indofine Chemical L-Abrine, 98%, 500 Mg
SDP
Supplier:  Indofine Chemical A001 500 MG
Encompass_Preferred
L-ABRINE 500 MG, Natural Products, N-Methyltryptophan, C12H14N2O2, MW: 218.25, >98% by HPLC, 526-31-8, MFCD00005645
Catalog No. 50-908-372