Learn More
TARGETMOL CHEMICALS INC Abrocitinib 10mg
SDP
Supplier:  TARGETMOL CHEMICALS INC TQ003710MG
Encompass_Preferred
Abrocitinib (PF-04965842) is a potent, specific and orally-active JAK1 inhibitor (IC50s: 29/803 nM for JAK1/2).
Catalog No. NC1910584