Learn More
Selleck Chemical LLC Abrocitinib2mg
SDP
Supplier:  Selleck Chemical LLC S87652MG
Encompass_Preferred
Abrocitinib (PF-04965842) is a potent JAK1 inhibitor with IC50s of 29 nM, 803 nM, > 10 000 nM and 1250 nM for JAK1, JAK2, JAK3 and tyrosine kinase (TYK) 2, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3388