Learn More
Grainger TapeCase Slick Surface Tape, PTFE Impregnated Fiberglass, 1.5 in x 36 yd, Silicone Adhesive, 1 ea
Catalog No. 502754995
Encompass_Preferred
This slick surface tape consists of a PTFE impregnated fiberglass backing coated with a high-temperature silicone adhesive. It is designed for demanding mechanical applications requiring a non-stick, abrasion-resistant release surface, such as heat sealing bars and conveyor belt coverings.
  • Features a durable PTFE coated fiberglass construction
  • Offers high temperature performance up to 260°C (500°F)
  • Suitable for use in assembly, manufacturing, and warehouse maintenance environments
  • Provides reliable release properties for heat sealers and form-fill-seal equipment
Catalog No. 50-275-4995 Supplier Grainger Supplier No. 15D386
This slick surface tape consists of a PTFE impregnated fiberglass backing coated with a high-temperature silicone adhesive. It is designed for demanding mechanical applications requiring a non-stick, abrasion-resistant release surface, such as heat sealing bars and conveyor belt coverings.
  • Features a durable PTFE coated fiberglass construction
  • Offers high temperature performance up to 260°C (500°F)
  • Suitable for use in assembly, manufacturing, and warehouse maintenance environments
  • Provides reliable release properties for heat sealers and form-fill-seal equipment