Learn More
Medchemexpress LLC Abrucomstat (3-Nitroxypropanol) | 100502-66-7 | 95.0% | 121.09 | 50 MG
SDP
Catalog No. 5000352133
Encompass_Preferred
Abrucomstat (3-Nitroxypropanol) is an orally active methyl coenzyme M reductase inhibitor that decreases ruminal methanogenesis. It is intended for research use only and is not sold to patients. The compound appears as a colorless to light yellow liquid with a density of 1.278±0.06 g/cm3.
  • Functions as an orally active methyl coenzyme M reductase inhibitor
  • Decreases ruminal methanogenesis
  • Reduces methane emissions in calves based on in vivo studies
  • Appears as a colorless to light yellow liquid
Catalog No. 50-003-52133 Supplier Medchemexpress LLC Supplier No. HY13956650MG
Abrucomstat (3-Nitroxypropanol) is an orally active methyl coenzyme M reductase inhibitor that decreases ruminal methanogenesis. It is intended for research use only and is not sold to patients. The compound appears as a colorless to light yellow liquid with a density of 1.278±0.06 g/cm3.
  • Functions as an orally active methyl coenzyme M reductase inhibitor
  • Decreases ruminal methanogenesis
  • Reduces methane emissions in calves based on in vivo studies
  • Appears as a colorless to light yellow liquid