Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Abrucomstat (3-Nitroxypropanol) | 100502-66-7 | MFCD31731012 | 95.0% | 121.09 | 100 MG
SDP

Catalog No. 5000351953
Encompass_Preferred

Abrucomstat (3-Nitroxypropanol) is an orally active methyl coenzyme M reductase inhibitor. It decreases ruminal methanogenesis and is intended for research use only.

  • Orally active methyl coenzyme M reductase inhibitor
  • Decreases ruminal methanogenesis
  • Intended for research use only
  • Store pure form at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO (100 mg/mL) and water (100 mg/mL)
  • Molecular formula: C3H7NO4
  • Molecular weight: 121.09
  • Purity: 95.0%

Catalog No. 50-003-51953 Supplier Medchemexpress LLC Supplier No. HY139566100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Abrucomstat (3-Nitroxypropanol) is an orally active methyl coenzyme M reductase inhibitor. It decreases ruminal methanogenesis and is intended for research use only.

  • Orally active methyl coenzyme M reductase inhibitor
  • Decreases ruminal methanogenesis
  • Intended for research use only
  • Store pure form at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO (100 mg/mL) and water (100 mg/mL)
  • Molecular formula: C3H7NO4
  • Molecular weight: 121.09
  • Purity: 95.0%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.