Learn More
American Biotech Supply Premier Undercounter Refrigerator Built-In - 4.6 Cu. Ft. Capacity
Catalog No. NC1653562
Encompass_Preferred
One glass swing door, right hinged. Microprocessor temperature controller. Digital temperature display. Audible and visual high and low temperature alarms. Remote alarm contacts. 2 probes; 1 in air and 1 in glycol bottle. Adjustable operating temperature range: 1°C – 10°C. Three shelves (two adjustable/one fixed) with guard rail on back to prevent product from pushing against back wall.
Catalog No. NC1653562 Supplier American Biotech Supply Supplier No. ABTHCUCBI0404G
One glass swing door, right hinged. Microprocessor temperature controller. Digital temperature display. Audible and visual high and low temperature alarms. Remote alarm contacts. 2 probes; 1 in air and 1 in glycol bottle. Adjustable operating temperature range: 1°C – 10°C. Three shelves (two adjustable/one fixed) with guard rail on back to prevent product from pushing against back wall.