Learn More
Waters Corp UV Detector Linearity Standards Kit for 3 mm Flow Cell
Catalog No. 50793623 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Kit contains standards appropriate for assessment of UV Detector with 3mm flow cell.7 Ampules (10 ml standard solution contains):Ampule 1 100% MethanolAmpule 2 15.0 mg/L Propyl Paraben (+/-1%)Ampule 3 30.0 mg/L Propyl Paraben (+/- 1%)Ampule 4 45.0 mg/L Propyl Paraben (+/- 1%)Ampule 5 60.0 mg/L Propyl Paraben (+/- 1%)Ampule 6 75.0 mg/L Propyl Paraben (+/- 1%)Ampule 7 90.0 mg/L Propyl Paraben (+/- 1%)Propyl Paraben (99% Purity).All standards are 100% HPLC Grade Methanol.
Catalog No. 50-793-623 Supplier Waters Corp Supplier No. WAT047634
Kit contains standards appropriate for assessment of UV Detector with 3mm flow cell.7 Ampules (10 ml standard solution contains):Ampule 1 100% MethanolAmpule 2 15.0 mg/L Propyl Paraben (+/-1%)Ampule 3 30.0 mg/L Propyl Paraben (+/- 1%)Ampule 4 45.0 mg/L Propyl Paraben (+/- 1%)Ampule 5 60.0 mg/L Propyl Paraben (+/- 1%)Ampule 6 75.0 mg/L Propyl Paraben (+/- 1%)Ampule 7 90.0 mg/L Propyl Paraben (+/- 1%)Propyl Paraben (99% Purity).All standards are 100% HPLC Grade Methanol.