Learn More
Fisherbrand™ MagLab™ MagBox™ Storage Box
GREENER_CHOICE
Catalog No. 01670379 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Translucent Orange
Catalog No. Color
01-670-379 Translucent Orange
1 options
Catalog No. 01-670-379
  • $31.00
    $24.65 / Each
    Save $6.35
    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $537.00
    $429.65 / Case of 20 EA
    Save $107.35
    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Create storage space without crowding limited workspace. Fisherbrand™ MagLab™ MagBox™ Storage Box uses magnetic mounting to store miscellaneous lab and office supplies off the benchtop. Constructed of durable, brightly colored ABS plastic.
Create lab storage space off your bench top with the MagLab™ storage system.
  • Easily organize common lab and office accessories off your bench top
  • Robust ABS plastic is translucent for easy content identification and access
  • Features four powerful magnets for a secure hold onto most metal surfaces
  • Mount on non-metallic surfaces with a self-adhesive metal mounting plate, provided
Specifications
Product Type Storage Container
Color Translucent Orange
Material ABS
Quantity 1 Ea.
Dimensions (L x W x H) 130 x 78 x 102 mm