Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Oatey ABS Cement, Medium-Bodied, Black, 16 oz., Fast Set

Catalog No. 502839679
Encompass

Oatey ABS Medium-Bodied Fast Set Black Cement is designed for non-pressure ABS pipe and fittings, up to 6 inches in diameter. Its black color aids application identification. The medium body provides excellent gap-filling properties and is easy to apply with the included in-lid dauber.

  • Solvent welds ABS pipe and fittings for non-pressure applications
  • Easy-to-apply medium body with excellent gap-filling properties
  • Suitable for all schedules and classes of ABS pipe and fittings up to 6 inches in diameter
  • Recommended application temperature: 40°F to 110°F (4°C to 43°C)
  • UL Greenguard Gold certified; low VOC, high-performance solvent cement
  • Meets ASTM Standard D2235 and complies with NSF/ANSI 61 health effects requirements

Catalog No. 50-283-9679 Supplier MSC Supplier No. 79747614
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oatey ABS Medium-Bodied Fast Set Black Cement is designed for non-pressure ABS pipe and fittings, up to 6 inches in diameter. Its black color aids application identification. The medium body provides excellent gap-filling properties and is easy to apply with the included in-lid dauber.

  • Solvent welds ABS pipe and fittings for non-pressure applications
  • Easy-to-apply medium body with excellent gap-filling properties
  • Suitable for all schedules and classes of ABS pipe and fittings up to 6 inches in diameter
  • Recommended application temperature: 40°F to 110°F (4°C to 43°C)
  • UL Greenguard Gold certified; low VOC, high-performance solvent cement
  • Meets ASTM Standard D2235 and complies with NSF/ANSI 61 health effects requirements

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.