Learn More
Instech ABS2 Dispensing Tip 20/18GA
SDP
Catalog No. NC3232501
Encompass
The Instech ABS2 Dispensing Tip is a replacement dispensing tip designed for use with the ABS2 Automated Blood Sampler. Engineered for precision transfer of blood and IV fluids, it supports accurate automated sampling workflows and integrates with large-animal tube set configurations for optimized flow and reliability.
Features
  • Type: Replacement dispensing tip for ABS2 automated blood sampler
  • Gauge: 18ga configuration for large-animal tube sets
  • Application: Supports automated blood dispensing and IV fluid return
  • Integration: Designed for ABS2 peristaltic pump pathways
  • Maintenance: Tip is serviceable and included in standard ABS2 maintenance routines
  • Quantity: Sold individually
Catalog No. NC3232501 Supplier Instech Supplier No. ABS2/ND2018
The Instech ABS2 Dispensing Tip is a replacement dispensing tip designed for use with the ABS2 Automated Blood Sampler. Engineered for precision transfer of blood and IV fluids, it supports accurate automated sampling workflows and integrates with large-animal tube set configurations for optimized flow and reliability.
Features
  • Type: Replacement dispensing tip for ABS2 automated blood sampler
  • Gauge: 18ga configuration for large-animal tube sets
  • Application: Supports automated blood dispensing and IV fluid return
  • Integration: Designed for ABS2 peristaltic pump pathways
  • Maintenance: Tip is serviceable and included in standard ABS2 maintenance routines
  • Quantity: Sold individually