Learn More
Cayman Chemical (+)-Abscisic Acid, 21293-29-8, 1 mg
SDP
Supplier:  Cayman Chemical 100731
Encompass_Preferred
Molecular Formula C15H20O4, Purity ≥98%, Formula Weight 264.3, Synonym: 5-[(1S)-1-hydroxy-2,6,6-trimethyl-4-oxo-2-cyclohexen-1-yl]-3-methyl-2Z,4E-pentadienoic acid; A plant hormone with diverse roles in disease resistance, plant development, and response to stresses; regulates gene expression and may contribute to epigenetic changes at the chromatin level.
Catalog No. NC1142174