Learn More
Medchemexpress LLC Abscisic acid | 21293-29-8 | MFCD00066545 | 264.32 g/mol | C15H20O4 | 1 ML
SDP
Catalog No. 5000213453
Encompass_Preferred
Abscisic acid is a naturally occurring phytohormone and research reagent used to study plant signaling, stress responses, and metabolic effects. This listing refers to the compound supplied as a 10 mM solution in a 1 mL vial for laboratory use; intended for research purposes only and not for human or clinical use.
  • Provides abscisic acid (ABA) as a research-grade reagent.
  • Supplied as a 10 mM solution in a 1 mL vial for convenient dosing.
  • Molecular formula C15H20O4; molecular weight 264.32 g/mol.
  • Suitable for in vitro and biochemical assays.
  • Not for human or clinical use; research use only.
  • Refer to the safety data sheet for handling and storage instructions.
Catalog No. 50-002-13453 Supplier Medchemexpress LLC Supplier No. HY10056010MM 1ML
Abscisic acid is a naturally occurring phytohormone and research reagent used to study plant signaling, stress responses, and metabolic effects. This listing refers to the compound supplied as a 10 mM solution in a 1 mL vial for laboratory use; intended for research purposes only and not for human or clinical use.
  • Provides abscisic acid (ABA) as a research-grade reagent.
  • Supplied as a 10 mM solution in a 1 mL vial for convenient dosing.
  • Molecular formula C15H20O4; molecular weight 264.32 g/mol.
  • Suitable for in vitro and biochemical assays.
  • Not for human or clinical use; research use only.
  • Refer to the safety data sheet for handling and storage instructions.