Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Abscisic acid | 21293-29-8 | MFCD00066545 | 99.7% | 264.32 | C15H20O4 | 100 MG
SDP

Catalog No. 5000213736
Encompass_Preferred

Abscisic acid is the (S)-(+)-enantiomer of the plant hormone supplied for research applications. It is used in plant biology and metabolic studies and has reported LANCL2 ligand and insulin-sensitizing activity. The compound is provided as a high-purity solid and in prepared solution formats for assay and analytical use.

  • High purity: 99.73%.
  • Molecular formula C15H20O4; molecular weight 264.32 g/mol.
  • CAS number 21293-29-8.
  • Available as solid and solution formats, including 100 MG quantities.
  • Suitable for plant biology, metabolic, and analytical research applications.
  • Provided with recommended solvent formats (10 mM in DMSO) for convenience.

Catalog No. 50-002-13736 Supplier Medchemexpress LLC Supplier No. HY100560100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Abscisic acid is the (S)-(+)-enantiomer of the plant hormone supplied for research applications. It is used in plant biology and metabolic studies and has reported LANCL2 ligand and insulin-sensitizing activity. The compound is provided as a high-purity solid and in prepared solution formats for assay and analytical use.

  • High purity: 99.73%.
  • Molecular formula C15H20O4; molecular weight 264.32 g/mol.
  • CAS number 21293-29-8.
  • Available as solid and solution formats, including 100 MG quantities.
  • Suitable for plant biology, metabolic, and analytical research applications.
  • Provided with recommended solvent formats (10 mM in DMSO) for convenience.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.