Learn More
Selleck Chemical LLC Abscisic Acid
SDP
Catalog No. 5000004132
Encompass_Preferred
Abscisic Acid (Dormin Abscisin II) is a plant hormone which is involved in many plant developmental processes modulates ion homeostasis and metabolism and inhibits germination and seedling growth
Catalog No. 50-000-04132 Supplier Selleck Chemical LLC Supplier No. S759425MG
Abscisic Acid (Dormin Abscisin II) is a plant hormone which is involved in many plant developmental processes modulates ion homeostasis and metabolism and inhibits germination and seedling growth