Learn More
TARGETMOL CHEMICALS INC ABSCISIC ACID 25MG
SDP
Catalog No. 502579411
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Abscisic Acid (Dormin) is a plant hormone which is involved in many plant developmental processes modulates ion homeostasis and metabolism and inhibits germination and seedling growth. purity: 98%
Catalog No. 50-257-9411 Supplier TARGETMOL CHEMICALS INC Supplier No. T636825MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Abscisic Acid (Dormin) is a plant hormone which is involved in many plant developmental processes modulates ion homeostasis and metabolism and inhibits germination and seedling growth. purity: 98%