Learn More
TARGETMOL CHEMICALS INC Abscisic acid 50MG
SDP
Catalog No. 502878916
Encompass_Preferred
Also available in 10 mg, 25 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. (±)-Abscisic acid is an orally active phytohormone also found in animals. It regulates blood glucose in mammals. Purity 99.87%
Catalog No. 50-287-8916 Supplier TARGETMOL CHEMICALS INC Supplier No. T301450MG
Also available in 10 mg, 25 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. (±)-Abscisic acid is an orally active phytohormone also found in animals. It regulates blood glucose in mammals. Purity 99.87%