Learn More
Apexbio Technology LLC Abscisic Acid (Dormin)(Synonyms: ABA, Dormin, Abscisin II, (+)-Abscisic acid, (S)-(+)-Abscisic acid, S-Abscisic acid, cis-Abscisic acid), 5mg, CAS: 21293-29-8.
SDP
Catalog No. 5000060992
Encompass_Preferred
Abscisic acid (Dormin CAS 21293-29-8) is a plant hormone involved in regulating plant growth development and stress responses Its activity includes mediating seed dormancy germination control lateral root formation and adaptive responses to abiotic stress Mechanistically ABA influences histone H2B ubiquitination and chromatin remodeling processes thereby modulating gene expression during seed maturation and under environmental stress ABA signaling also alters cellular redox status by enhancing synthesis of antioxidants such as -tocopherol and L-ascorbate It is widely utilized in plant biology research to study stress physiology developmental biology and epigenetic gene regulation
Catalog No. 50-000-60992 Supplier Apexbio Technology LLC Supplier No. B48835
Abscisic acid (Dormin CAS 21293-29-8) is a plant hormone involved in regulating plant growth development and stress responses Its activity includes mediating seed dormancy germination control lateral root formation and adaptive responses to abiotic stress Mechanistically ABA influences histone H2B ubiquitination and chromatin remodeling processes thereby modulating gene expression during seed maturation and under environmental stress ABA signaling also alters cellular redox status by enhancing synthesis of antioxidants such as -tocopherol and L-ascorbate It is widely utilized in plant biology research to study stress physiology developmental biology and epigenetic gene regulation