Learn More
Cayman Chemical (+)-Abscisic Acid, Purity: ≥98%, CAS: 21293-29-8, 50 mg
SDP
Catalog No. 502891384
Encompass_Preferred
A plant hormone with diverse roles in disease resistance, plant development, and response to stresses; regulates gene expression and may contribute to epigenetic changes at the chromatin level
Catalog No. 50-289-1384 Supplier Cayman Chemical Supplier No. 1007350 MG
A plant hormone with diverse roles in disease resistance, plant development, and response to stresses; regulates gene expression and may contribute to epigenetic changes at the chromatin level