Learn More
Medchemexpress LLC (+)-Abscisic aldehyde | 41944-86-9 | 248.32 g/mol | C15H20O3 | 1 MG
SDP
Catalog No. 5000284063
Encompass_Preferred
(+)-Abscisic aldehyde is the aldehyde precursor of the plant hormone abscisic acid (ABA). It is used as a biochemical reagent for research into ABA biosynthesis, metabolism, and plant physiological responses, and is suitable for analytical and enzymology studies.
  • Used to study abscisic acid biosynthesis and metabolism.
  • Applicable in plant physiology, stress-response, and enzymology research.
  • Chemical formula C15H20O3; molecular weight 248.32 g/mol.
  • Provided in small research quantities suitable for analytical and preparative experiments.
  • Handle and store according to the certificate of analysis and safety data sheet.
Catalog No. 50-002-84063 Supplier Medchemexpress LLC Supplier No. HYW6537391MG
(+)-Abscisic aldehyde is the aldehyde precursor of the plant hormone abscisic acid (ABA). It is used as a biochemical reagent for research into ABA biosynthesis, metabolism, and plant physiological responses, and is suitable for analytical and enzymology studies.
  • Used to study abscisic acid biosynthesis and metabolism.
  • Applicable in plant physiology, stress-response, and enzymology research.
  • Chemical formula C15H20O3; molecular weight 248.32 g/mol.
  • Provided in small research quantities suitable for analytical and preparative experiments.
  • Handle and store according to the certificate of analysis and safety data sheet.