Learn More
BD™ AbSeq Immune Discovery Panel
Catalog No. BDB625970 Shop All BD Biosciences Products
Change view
Click to view available options
For Use With (Equipment):
BD Rhapsody Platform, Single-Cell Analysis System
Catalog No. For Use With (Equipment)
BDB625970 BD Rhapsody Platform, Single-Cell Analysis System
1 options
Catalog No. BDB625970 Supplier BD™ Supplier No. 625970
Add to Cart
Add to Cart
BD AbSeq Immune Discovery Panel is developed using BD antibody-oligo based technology and consists of 30 different specificities against major human immune markers lyophilized in a single tube.
Performing protein discovery experiments can be challenging. From reagent handling and marker selection to ensuring accurate execution of all the necessary steps, probing samples for tens of protein markers in a single experiment is no trivial task. Introducing the BD™ AbSeq Immune Discovery Panel (IDP), a simple tool that will enable you to uncover up to 30 different immune markers in a single experiment.
  • The IDP consists of 30 different specificities against major human immune markers combined in a single tube.
  • Developed to ensure IDP performance was comparable to data generated by freshly prepared mixture of the same 30 specificities of BD single vial BD™ AbSeq Reagents.
  • Designed and optimized to work on the BD Rhapsody™ HT Single-Cell Analysis System
  • Tested to work seamlessly with BD Rhapsody™ Single-Cell RNA Assays and Multiplexing Kits.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) BD Rhapsody Platform, Single-Cell Analysis System
Product Type Antiobody-Oligonuceltide Conjugates