Learn More
MSC Igaging Lcd Digital Drop Indicator, 4 in. Range, 0.0004 in. Accuracy, IP67, 1 ea.
Catalog No. 5000510670
Encompass
This LCD digital electronic drop indicator provides high-precision measurements up to 4 in. (101 mm) with an accuracy of 0.0004 in. (0.01 mm). Designed for robust performance in industrial environments, this device features a durable ceramic contact point and an IP67 rating for protection against dust and water. It includes both flat and lug back options for versatile mounting and supports smart wireless data output for efficient measurement recording.
  • Measurement range of 0 to 4 in. (101 mm)
  • High-resolution display with 0.00005 in. (0.001 mm) resolution
  • IP67 water and dust protective rating
  • Ceramic contact point
  • Smart wireless data output and in/mm reading tolerance
  • Includes case, battery, and inspection report with declaration of conformity
Catalog No. 50-005-10670 Supplier MSC Supplier No. 84648864
This LCD digital electronic drop indicator provides high-precision measurements up to 4 in. (101 mm) with an accuracy of 0.0004 in. (0.01 mm). Designed for robust performance in industrial environments, this device features a durable ceramic contact point and an IP67 rating for protection against dust and water. It includes both flat and lug back options for versatile mounting and supports smart wireless data output for efficient measurement recording.
  • Measurement range of 0 to 4 in. (101 mm)
  • High-resolution display with 0.00005 in. (0.001 mm) resolution
  • IP67 water and dust protective rating
  • Ceramic contact point
  • Smart wireless data output and in/mm reading tolerance
  • Includes case, battery, and inspection report with declaration of conformity