Learn More
Agilent Technologies ABSOLUTELY RNA MICROPREP KITA
Catalog No. 50125670
Encompass
Absolutely RNA Microprep Kit isolate total RNA from up to 5 x 105 cells. Efficient on-column DNA removal. Contains RNA binding spin cups, lysis buffer, beta-mercaptoethanol, DNase (lyophilized), DNase buffers, wash buffers, elution buffer
Catalog No. 50-125-670 Supplier Agilent Technologies Supplier No. 400805
Absolutely RNA Microprep Kit isolate total RNA from up to 5 x 105 cells. Efficient on-column DNA removal. Contains RNA binding spin cups, lysis buffer, beta-mercaptoethanol, DNase (lyophilized), DNase buffers, wash buffers, elution buffer