Learn More
Agilent Technologies ABSOLUTELY RNA NANOPREP KITAB
Catalog No. 50125656
Encompass
Absolutely RNA Nanoprep Kit 50 preps The Absolutely RNA nanoprep kit offers easy purification of DNA-free total RNA from the smallest samples eluted in a volume of 10microliter to provide RNA at a concentration useful for many downstream ap
Catalog No. 50-125-656 Supplier Agilent Technologies Supplier No. 400753
Absolutely RNA Nanoprep Kit 50 preps The Absolutely RNA nanoprep kit offers easy purification of DNA-free total RNA from the smallest samples eluted in a volume of 10microliter to provide RNA at a concentration useful for many downstream ap