Learn More
Sigma Aldrich Fine Chemicals Biosciences MiniPax absorbent packets, molecular sieves, weight 1 g (desiccant), L × W 3/4 in. × 1 7/16 in.
Catalog No. NC1539231
Encompass
MiniPax packets are compact, extremely durable, non-dusting, and non-linting with high water vapor trasmission rate and controlled porosity. The packets take up little space in packaging and have a high moisture adsorbing capacity. Envelopes are formed from sheet of polyethylene fibers has high wet strength and is permeable to both gases and odors. Indicating MiniPax contain cobalt chloride and should not be used for food or pharmaceutical applications.
Catalog No. NC1539231 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z163619100EA
MiniPax packets are compact, extremely durable, non-dusting, and non-linting with high water vapor trasmission rate and controlled porosity. The packets take up little space in packaging and have a high moisture adsorbing capacity. Envelopes are formed from sheet of polyethylene fibers has high wet strength and is permeable to both gases and odors. Indicating MiniPax contain cobalt chloride and should not be used for food or pharmaceutical applications.