Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Absorb Detector Cell Solution, 10 mm, Performance Qualification, UV System Assessment

Catalog No. 50466411 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This product is a kit containing standards appropriate for the assessment of HPLC UV systems. This solution is specifically designed for performance qualification of absorb detectors.

  • Contains standards for HPLC UV system assessment.
  • Liquid formulation for UV detection.
  • Suitable for LC system types.
  • Quantitative analysis capabilities.
  • Shelf life of 24 months.
  • Stores at room temperature.
  • Ships under ambient conditions.
  • Reference type product.
  • Each ampoule contains 10 mL of standard solution.
  • Each package contains 1 unit.
  • Standard solution comprises 10.00 mg/L Acetophenone (purity > 99%), 10.00 mg/L Butyrophenone (purity > 99%), and 3.2 g/L Acetone (purity > 99.74%).
  • Solution in 60% HPLC Grade Methanol / 40% HPLC Grade Water, volume/volume.
  • Product is considered hazardous material, requiring special freight handling.

Catalog No. 50-466-411 Supplier Waters Corp Supplier No. 186006684
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is a kit containing standards appropriate for the assessment of HPLC UV systems. This solution is specifically designed for performance qualification of absorb detectors.

  • Contains standards for HPLC UV system assessment.
  • Liquid formulation for UV detection.
  • Suitable for LC system types.
  • Quantitative analysis capabilities.
  • Shelf life of 24 months.
  • Stores at room temperature.
  • Ships under ambient conditions.
  • Reference type product.
  • Each ampoule contains 10 mL of standard solution.
  • Each package contains 1 unit.
  • Standard solution comprises 10.00 mg/L Acetophenone (purity > 99%), 10.00 mg/L Butyrophenone (purity > 99%), and 3.2 g/L Acetone (purity > 99.74%).
  • Solution in 60% HPLC Grade Methanol / 40% HPLC Grade Water, volume/volume.
  • Product is considered hazardous material, requiring special freight handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.