Learn More
Enzo Life Sciences Enzo Life Sciences Absorbance 96 Plate Reader, Compact Design, USB Powered, 96 Detection Units
SDP
Catalog No. NC3653536
Encompass_Preferred
The Enzo Life Sciences Absorbance 96 Plate Reader offers a compact solution for various scientific applications, such as ELISA and protein quantification. Its small footprint saves benchtop space while its solid-state design ensures maintenance-free operation and rapid measurements. With a USB power supply and user-friendly software compatible with both Windows and macOS, it delivers precise and accurate results at an affordable price.
Catalog No. NC3653536 Supplier Enzo Life Sciences Supplier No. ENZINSA96
Add to Cart
Add to Cart
Request a Quote
The Enzo Life Sciences Absorbance 96 Plate Reader offers a compact solution for various scientific applications, such as ELISA and protein quantification. Its small footprint saves benchtop space while its solid-state design ensures maintenance-free operation and rapid measurements. With a USB power supply and user-friendly software compatible with both Windows and macOS, it delivers precise and accurate results at an affordable price.