Learn More
Chemglass Life Sciences Absorbent Bench-Top Liner, 20" x 50'
SDP
Catalog No. 501897542
Encompass
Absorbent Bench-Top Liner, 20" x 50'. Absorbent liner keeps spills from spreading and protects benchtops, floors, and carts. Cushions surfaces while absorbing liquids. Made of highly absorbent three-ply tissue with a waterproof polypropylene backing that is resistant to most chemicals.
Catalog No. 50-189-7542 Supplier Chemglass Life Sciences Supplier No. CGE1150001
Absorbent Bench-Top Liner, 20" x 50'. Absorbent liner keeps spills from spreading and protects benchtops, floors, and carts. Cushions surfaces while absorbing liquids. Made of highly absorbent three-ply tissue with a waterproof polypropylene backing that is resistant to most chemicals.