Learn More
Bachem H-Pro-Pro-Asp-NH2
SDP
Supplier:  Bachem 4090413.1000
Encompass_Preferred
H-Pro-Pro-Asp-NH2 Trifluoroacetate, 850440-85-6, C14H22N4O5, Mr 326.35, 1g. Absorbed to modified silica gel, PPD-amide is a highly efficient recyclable chiral catalyst for aldol reactions.
Catalog No. 50-194-6873