Learn More
Alkali Scientific ASI™ Absorbent Bench-Top Liner, 20" Wide, 24" Sheet, 50/Case
SDP
Catalog No. 503026552
Encompass_Preferred
Protect surfaces from spills with economical, highly absorbent. bench liners that allow for safe, easy cleanup Soft, durable, and highly absorbent multilayer liner cushions surfaces and absorbs and retains spills Waterproof polypropylene backing resists most chemicals Easily cut to desired size White Ideal for protecting benches and surfaces Available in roll or dispenser box
Catalog No. 50-302-6552 Supplier Alkali Scientific Supplier No. ABL24
Protect surfaces from spills with economical, highly absorbent. bench liners that allow for safe, easy cleanup Soft, durable, and highly absorbent multilayer liner cushions surfaces and absorbs and retains spills Waterproof polypropylene backing resists most chemicals Easily cut to desired size White Ideal for protecting benches and surfaces Available in roll or dispenser box