Learn More
Andwin Scientific ABSORBENT PAPER 18X20 400
SDP
Catalog No. NC9205028
Encompass_Preferred
The Absorbent Laboratory Countertop Paper is constructed with thick, absorbent double re-crepe (DRC) material that gives a soft, cloth-like feel. This product is pebble embossed for extra cushioning. Its backing is poly-coated to have a fluid holdout. The flat sheet measures 45.7W x 50.8L cm (18 x 20). It is sold in a case of 400.
Catalog No. NC9205028 Supplier Andwin Scientific Supplier No. 54410530
The Absorbent Laboratory Countertop Paper is constructed with thick, absorbent double re-crepe (DRC) material that gives a soft, cloth-like feel. This product is pebble embossed for extra cushioning. Its backing is poly-coated to have a fluid holdout. The flat sheet measures 45.7W x 50.8L cm (18 x 20). It is sold in a case of 400.