Learn More
Enterprise Technology Solutions Absorbent Specialty Products Dams,f/Floodwater,Gel Interior,10'Lx2.5"H,12/CT,Orange
SDP
Catalog No. 5000790269
Encompass_Preferred
Water Dams quickly absorb and gel water, swelling to create a barrier to control leaks and spills. Compact, water-activated dams swell and gel when wet. They are lightweight, quick and easy to deploy. Each one expands to 2.5" high in minutes to contain, control and divert problem water. Long, continuous lengths are perfect for wraparound protection. Durable knit keeps inner gel contained. Water dams can be left in place for maintenance or discarded into regular trash.
Catalog No. 50-007-90269 Supplier Enterprise Technology Solutions Supplier No. APRWUB21012
Water Dams quickly absorb and gel water, swelling to create a barrier to control leaks and spills. Compact, water-activated dams swell and gel when wet. They are lightweight, quick and easy to deploy. Each one expands to 2.5" high in minutes to contain, control and divert problem water. Long, continuous lengths are perfect for wraparound protection. Durable knit keeps inner gel contained. Water dams can be left in place for maintenance or discarded into regular trash.