Learn More
Enterprise Technology Solutions Absorbent Specialty Products Flood Bags,4-Gal Capacity,3-1/2 Expansion,12"x24",20/CT,BK
SDP
Catalog No. 5000790505
Encompass_Preferred
Flood Bags absorb, contain and divert problem floodwater to help protect your property from the dangers of flooding. They activate when wet and require no sand. Each bag absorbs 4 gallons of water, swells to 32 lb. and expands up to 3-1/5" high in 10 minutes. This compact and lightweight flood control is quick and easy to deploy. Inner absorbent turns to gel and creates a barrier. Flat uniform shape allows easy stacking. Stack in brick and pyramid formation. Leave bags in place for ongoing protection. No additional tools or supplies are needed to set up this type of flood control. Bags are safe, nontoxic and nonhazardous. When you no longer need them, you can discards them into your regular trash.
Catalog No. 50-007-90505 Supplier Enterprise Technology Solutions Supplier No. APRQD122420
Flood Bags absorb, contain and divert problem floodwater to help protect your property from the dangers of flooding. They activate when wet and require no sand. Each bag absorbs 4 gallons of water, swells to 32 lb. and expands up to 3-1/5" high in 10 minutes. This compact and lightweight flood control is quick and easy to deploy. Inner absorbent turns to gel and creates a barrier. Flat uniform shape allows easy stacking. Stack in brick and pyramid formation. Leave bags in place for ongoing protection. No additional tools or supplies are needed to set up this type of flood control. Bags are safe, nontoxic and nonhazardous. When you no longer need them, you can discards them into your regular trash.