Learn More
Lonza Walkersville Ascorbic Acid, 0.5 mL
Catalog No. NC1310079
Encompass
Ascorbic Acid is an important component in various primary cell culture media kits. It is included in various BulletKits. Ascorbic Acid is also sold separately at a concentration of 70 mM.Content:1 x Vial Containing 70 mM Ascorbic Acid 0.5 mL
Catalog No. NC1310079 Supplier Lonza Walkersville Supplier No. CC4398
Add to Cart
Add to Cart
Ascorbic Acid is an important component in various primary cell culture media kits. It is included in various BulletKits. Ascorbic Acid is also sold separately at a concentration of 70 mM.Content:1 x Vial Containing 70 mM Ascorbic Acid 0.5 mL