Learn More
Research Products International Corp AbsorbMax Black Sealing Film, Non-Sterile, 50 per Package
SDP
Catalog No. 50998792
Encompass
AbsorbMax black sealing film provides a quick and convenient method for protecting light sensitive samples contained in 96 well plates. This light absorbing film reduces stray light and crosstalk in fluorescence assays. Fits within rim of raised rim plates and can be applied to either top or bottom of plate. AbsorbMax is a Trademark of Excel Scientific, Inc.
Catalog No. 50-998-792 Supplier Research Products International Corp Supplier No. 202537
AbsorbMax black sealing film provides a quick and convenient method for protecting light sensitive samples contained in 96 well plates. This light absorbing film reduces stray light and crosstalk in fluorescence assays. Fits within rim of raised rim plates and can be applied to either top or bottom of plate. AbsorbMax is a Trademark of Excel Scientific, Inc.