Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Absorption Quartz Micro Rectangular Cells with PTFE Lid
SCI_ED

Catalog No. S35864 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S35864 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S35864

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fit standard spectrophotometer cell holders

  • All cells are 12.5mm wide
  • 10mm path length rectangular cells
  • Reduce reagent use with Micro cells
  • Quartz windows for transmission from far ultraviolet through the infrared

Specifications

Type Micro Absorption Cell
Capacity (Metric) 0.5 mL
Material Quartz
Height (Metric) 45 mm
Width (Metric) 2 mm
For Use With (Application) UV/Vis Spectrophotometry
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.