Learn More
Innovating Science™ Absorption Spectroscopy Demonstration
SCI_ED
Catalog No. S01999 Shop All Aldon Products
Change view
Click to view available options
Grade:
6+
Catalog No. Grade
S01999 6+
1 options
Catalog No. S01999 Supplier Innovating Science™ Supplier No. IS7051
Add to Cart
Add to Cart
Explore the properties of atomic absorption and discover how certain elements absorb several specific wavelengths of light by viewing the dark line (absorption) spectra of various colored solutions including solutions containing elements that astronomers observe: erbium and praseodymium.
Explore the properties of atomic absorption and discover how certain elements absorb several specific wavelengths of light by viewing the dark line (absorption) spectra of various colored solutions including solutions containing elements that astronomers observe: erbium and praseodymium.
Specifications
Grade 6+
Product Type Physics Demonstration