Learn More
Medchemexpress LLC HY-15197 5mg Medchemexpress, ABT-046 CAS:1031336-60-3 Purity:>98%
SDP
Catalog No. 502081692
Encompass_Preferred
Medchemexpress, HY-15197 5mg ABT-046 CAS:1031336-60-3 ABT-046 is a potent, selective, and orally efficacious acyl CoA:diacylglycerol acyltransferase 1 (DGAT-1) inhibitor (IC50= 8 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1692 Supplier Medchemexpress LLC Supplier No. HY151975MG
Medchemexpress, HY-15197 5mg ABT-046 CAS:1031336-60-3 ABT-046 is a potent, selective, and orally efficacious acyl CoA:diacylglycerol acyltransferase 1 (DGAT-1) inhibitor (IC50= 8 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.