Learn More
Medchemexpress LLC HY-101634 5mg Medchemexpress, ABT-072 CAS:1132936-00-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1016345MG
Encompass_Preferred
Medchemexpress, HY-101634 5mg ABT-072 CAS:1132936-00-5 ABT-072 is a nonnucleoside NS5B polymerase inhibitor and a candidate drug evaluated for treatment of hepatitis C virus. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2185