Learn More
Medchemexpress LLC Abt-072 (potassium trihydrate) | 1132940-31-8 | MFCD07782111 | 99.6% | 561.69 g/mol | C24H32KN3O8S | 25 MG
SDP
Catalog No. 5000245477
Encompass_Preferred
ABT-072 (potassium trihydrate) is an orally active, potent non-nucleoside hepatitis C virus (HCV) NS5B polymerase inhibitor with subnanomolar activity against HCV genotypes 1a and 1b. Supplied for research use as the potassium trihydrate salt; molecular formula C24H32KN3O8S, molecular weight 561.69 g/mol.
  • Oral non-nucleoside NS5B polymerase inhibitor
  • Subnanomolar potency (HCV GT1a EC50 ≈ 1 nM; GT1b EC50 ≈ 0.3 nM)
  • High purity (~99.6%)
  • Provided as potassium trihydrate salt
  • Recommended storage: 4°C sealed; in solution -80°C up to 6 months, -20°C up to 1 month
  • Not classified as hazardous under GHS
Catalog No. 50-002-45477 Supplier Medchemexpress LLC Supplier No. HY101634A25MG
ABT-072 (potassium trihydrate) is an orally active, potent non-nucleoside hepatitis C virus (HCV) NS5B polymerase inhibitor with subnanomolar activity against HCV genotypes 1a and 1b. Supplied for research use as the potassium trihydrate salt; molecular formula C24H32KN3O8S, molecular weight 561.69 g/mol.
  • Oral non-nucleoside NS5B polymerase inhibitor
  • Subnanomolar potency (HCV GT1a EC50 ≈ 1 nM; GT1b EC50 ≈ 0.3 nM)
  • High purity (~99.6%)
  • Provided as potassium trihydrate salt
  • Recommended storage: 4°C sealed; in solution -80°C up to 6 months, -20°C up to 1 month
  • Not classified as hazardous under GHS