Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Abt-072 (potassium trihydrate) | 1132940-31-8 | MFCD34567113 | 99.6% | 561.69 g/mol | C24H32KN3O8S | 10 MG
SDP

Catalog No. 5000249104
Encompass_Preferred

ABT-072 (potassium trihydrate) is the potassium salt trihydrate form of a non-nucleoside hepatitis C virus (HCV) NS5B polymerase inhibitor supplied for research use. It shows nanomolar in vitro potency (HCV GT1a EC50 = 1 nM; GT1b EC50 = 0.3 nM) and is identified by CAS 1132940-31-8.

  • High chemical purity suitable for research (99.6%).
  • Potassium salt trihydrate form to improve solubility and handling.
  • Nanomolar inhibitory activity against HCV NS5B polymerase.
  • Available in small research sizes (1 mg, 5 mg, 10 mg, 25 mg, 50 mg).
  • Stable when sealed at 4°C; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Formula: C24H32KN3O8S; molecular weight: 561.69 g/mol.

Catalog No. 50-002-49104 Supplier Medchemexpress LLC Supplier No. HY101634A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ABT-072 (potassium trihydrate) is the potassium salt trihydrate form of a non-nucleoside hepatitis C virus (HCV) NS5B polymerase inhibitor supplied for research use. It shows nanomolar in vitro potency (HCV GT1a EC50 = 1 nM; GT1b EC50 = 0.3 nM) and is identified by CAS 1132940-31-8.

  • High chemical purity suitable for research (99.6%).
  • Potassium salt trihydrate form to improve solubility and handling.
  • Nanomolar inhibitory activity against HCV NS5B polymerase.
  • Available in small research sizes (1 mg, 5 mg, 10 mg, 25 mg, 50 mg).
  • Stable when sealed at 4°C; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Formula: C24H32KN3O8S; molecular weight: 561.69 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.