Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Abt-072 potassium trihydrate | 1132940-31-8 | MFCD34567113 | 99.6% | 561.69 g·mol⁻¹ | C24H32KN3O8S | 5 MG
SDP

Catalog No. 5000248586
Encompass_Preferred

ABT-072 potassium trihydrate is an orally active, potent non-nucleoside hepatitis C virus (HCV) NS5B polymerase inhibitor with nanomolar activity against genotype 1a and 1b. Supplied as a potassium salt trihydrate in powder form for research use only. Typical handling: store powder at -20 °C; solutions in DMSO are stable short-term at 4 °C and long-term at -80 °C.

  • Potent NS5B polymerase inhibition (HCV GT1a EC50 = 1 nM; GT1b EC50 = 0.3 nM).
  • Potassium trihydrate salt supplied as a powder.
  • High purity suitable for biochemical and virology research.
  • CAS RN 1132940-31-8 for unambiguous identification.
  • Recommended storage: -20 °C (powder); 4 °C short-term or -80 °C long-term in DMSO.

Catalog No. 50-002-48586 Supplier Medchemexpress LLC Supplier No. HY101634A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ABT-072 potassium trihydrate is an orally active, potent non-nucleoside hepatitis C virus (HCV) NS5B polymerase inhibitor with nanomolar activity against genotype 1a and 1b. Supplied as a potassium salt trihydrate in powder form for research use only. Typical handling: store powder at -20 °C; solutions in DMSO are stable short-term at 4 °C and long-term at -80 °C.

  • Potent NS5B polymerase inhibition (HCV GT1a EC50 = 1 nM; GT1b EC50 = 0.3 nM).
  • Potassium trihydrate salt supplied as a powder.
  • High purity suitable for biochemical and virology research.
  • CAS RN 1132940-31-8 for unambiguous identification.
  • Recommended storage: -20 °C (powder); 4 °C short-term or -80 °C long-term in DMSO.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.