Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ABT-072 (potassium trihydrate) | 1132940-31-8 | MFCD34567113 | 99.6% | 561.69 g/mol | C24H32KN3O8S | 50 MG
SDP

Catalog No. 5000248692
Encompass_Preferred

ABT-072 (potassium trihydrate) is an orally active, non-nucleoside inhibitor of the hepatitis C virus NS5B RNA-dependent RNA polymerase, provided as the potassium trihydrate salt for research and analytical applications. It exhibits potent nanomolar activity against genotype 1 HCV polymerases and is supplied in small lab pack sizes suitable for screening and characterization.

  • Orally active non-nucleoside NS5B polymerase inhibitor.
  • Nanomolar potency against HCV genotype 1 (EC50 ≈ 1 nM for GT1a, 0.3 nM for GT1b).
  • High purity suitable for analytical and biochemical studies.
  • Molecular formula C24H32KN3O8S and molecular weight 561.69 g/mol.
  • Supplied as a potassium trihydrate salt for improved handling and stability.
  • Packaged in small quantities for research screening and characterization.

Catalog No. 50-002-48692 Supplier Medchemexpress LLC Supplier No. HY101634A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ABT-072 (potassium trihydrate) is an orally active, non-nucleoside inhibitor of the hepatitis C virus NS5B RNA-dependent RNA polymerase, provided as the potassium trihydrate salt for research and analytical applications. It exhibits potent nanomolar activity against genotype 1 HCV polymerases and is supplied in small lab pack sizes suitable for screening and characterization.

  • Orally active non-nucleoside NS5B polymerase inhibitor.
  • Nanomolar potency against HCV genotype 1 (EC50 ≈ 1 nM for GT1a, 0.3 nM for GT1b).
  • High purity suitable for analytical and biochemical studies.
  • Molecular formula C24H32KN3O8S and molecular weight 561.69 g/mol.
  • Supplied as a potassium trihydrate salt for improved handling and stability.
  • Packaged in small quantities for research screening and characterization.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.