Learn More
Selleck Chemical LLC Venetoclax (ABT-199) 50mg 1257044-40-8 GDC-0199
SDP
Supplier:  Selleck Chemical LLC S804850MG
Encompass_Preferred
Venetoclax (ABT-199, GDC-0199) is a Bcl-2-selective inhibitor with Ki of <0.01 nM in cell-free assays, >4800-fold more selective versus Bcl-xL and Bcl-w, and no activity to Mcl-1. Venetoclax is reported to induce cell growth suppression, apoptosis, cell cycle arrest, and autophagy in triple negative breast cancer MDA-MB-231 cells. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0563538